missing translation for 'onlineSavingsMsg'
Learn More

Estramustine phosphate sodium, MedChemExpress

MedChemExpress Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.

95.85 € - 222.75 €

Specifications

Chemical Name or Material Estramustine phosphate sodium
CAS 52205-73-9
Color White
Purity Grade Notes Research
Molecular Formula C23H30Cl2NNa2O6P
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475778
Encompass_Preferred
View Documents
Medchem Express
HY-13627
10 mg
95.85 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16451937
Encompass_Preferred
View Documents
Medchem Express
HY-13627
50 mg
157.95 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16426256
Encompass_Preferred
View Documents
Medchem Express
HY-13627
100 mg
222.75 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.
Specifications

Specifications

Estramustine phosphate sodium
White
C23H30Cl2NNa2O6P
O=C(N(CCCl)CCCl)OC1=CC2=C(C=C1)[C@@]3([H])CC[C@@]4(C)[C@](CC[C@@H]4OP(O[Na])(O[Na])=O)([H])[C@]3([H])CC2
564.35
Research
Cancer-programmed cell death
52205-73-9
Research
H2O : 62.5 mg/mL (110.75 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.86 mM; Need ultrasonic)
564.35
96.79%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)

Recommended Storage : 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)

SDS
Documents

Documents

Product Certifications

Research purposes only