Learn More
Estrogen Receptor Ab-11 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Estrogen Receptor Ab-11, Mouse Monoclonal Antibody.
214.20 € - 1340.00 €
Specifications
Antigen | Estrogen Receptor Ab-11 |
---|---|
Clone | 1D5 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12621137
|
Epredia
MS-354-P0 |
Concentrated | 100 μL | N/A | |||||
12651137
|
Epredia
MS-354-P1ABX |
Concentrated | 100 μL | N/A | |||||
12671137
|
Epredia
MS-354-PABX |
Concentrated | 200 μL | N/A | |||||
12641137
|
Epredia
MS-354-P1 |
Concentrated | 500 μL |
779.00 €
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12631137
|
Epredia
MS-354-P |
Concentrated | 1 mL |
1245.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12601147
|
Epredia
MS-354-R7 |
Prediluted | 7 mL | N/A | |||||
Description
The ER gene consists of more than 140kb of genomic DNA divided into 8 exons and is translated into a protein with six functionally discrete domains required for transcription activation function, binding to estrogen response element (ERE) constitutive dimerization, binding to heat shock proteins, and ligand recognition. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.Host Species: Mouse
Clone: 1D5
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: N-terminal domain
Immunogen: Recombinant human estrogen receptor protein
Molecular Weight: 67 kDa
Positive Control: MCF-7 cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Estrogen Receptor Ab-11 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ASR, IVD | |
Human, Mouse, Non-human Primate, Rat | |
IgG2a κ |
1D5 | |
Monoclonal | |
Mouse | |
Steroids Hormones and Growth Factors | |
Recombinant human estrogen receptor protein | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.