Learn More
IKA™ ETH SER 8 Ethernet-Serial Converter
With the ETH SER 8 Ethernet-Serial Converter, up to eight laboratory devices can be controlled or read out via ethernet. The converter is configured using a separate PC software.
Brand: IKA™ 0020024286
This item is not returnable.
View return policy
Description
- Ports support RS-232, RS-422, and RS-485
- Converter is configured using a separate PC software
- Includes power cord, ethernet cable, wall mounting kit, and software CD
Specifications
Ethernet-serial Converter | |
RS-232 / RS-422 / RS-485 (8 Ports) | |
Power Cord, Ethernet Cable, Wall Mounting Kit, and Software CD |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
IKA™ ETH SER 8 Ethernet-Serial Converter > Ethernet-Serial Converter; Connector Types: RS-232 / RS-422 / RS-485 (8 Ports)
Spot an opportunity for improvement?Share a Content Correction