missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Pico Rabbit IgG Detection Kit

Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.

Brand:  Thermo Scientific™ 34083

Product Code. 10590814

  • 497.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.

All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.

Highlights:

  • Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer

  • Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates

  • Instructions – complete, detailed manual with recommended antibody dilutions

Ordering Information:

Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.

Includes:

Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SuperSignal West Pico Rabbit IgG Detection Kit
Chemiluminescence
Nitrocellulose, PVDF
500 mL
Horseradish Peroxidase
50 mini blots (4000 sq. cm of membrane)
Rabbit IgG Detection Kit
Western Blot
SuperSignal™
HRP
Rabbit
Sufficient For: ∼62 mini blots probed with rabbit primary antibodies
• SuperSignal West Pico Luminol/Enhancer Solution, 250 mL
• SuperSignal West Pico Stable Peroxide Solution, 250 mL
• Peroxidase Conjugated Goat anti-Rabbit IgG (H + L), 2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZF382876 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number34083
Lot NumberYE364963 Certificate TypeCertificate of Analysis Date15/05/2023 Catalog Number34083
Lot NumberXH362333 Certificate TypeCertificate of Analysis Date03/09/2022 Catalog Number34083
Lot NumberWF325550 Certificate TypeCertificate of Analysis Date09/06/2021 Catalog Number34083
Lot NumberVB295366 Certificate TypeCertificate of Analysis Date04/03/2020 Catalog Number34083
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SuperSignal™ West Pico Rabbit IgG Detection Kit > Rabbit SuperSignal kit; 500mL kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.