missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 99.5%, BAKER HPLC ANALYZED, J.T. Baker™ Product Code.: 10538071
compliance-icons compliance-icons

Ethanol, 99.5%, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 10538071
2.5 L, Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 10538071

Brand: JT Baker 8462.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

HPLC grade ethanol for use in High Performance Liquid Chromatography, packaged in 2.5L glass bottles.

BAKER ANALYZED™ HPLC grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Manufactured and tested to assure interference-free separations
  • Highly characterized reagents that have unmatched lot-to-lot consistency
  • Minimizes recalibration or instrument adjustments
  • Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
  • An Actual Lot Analysis is provided on the label

Specifications

Melting Point −114.5°C
Absorbance Max. 0.7 at 210nm, Max. 0.1 at 240nm, Max. 0.01 at 260nm
CAS 64-17-5
Color Colorless
Density 0.79g/cm3
Boiling Point 78°C
Quantity 2.5 L
Explosion Limit Lower: 3.5 Vol %, Upper: 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Molecular Formula C2H5OH
Viscosity 1.2 mPas at 20°C
Water Max. 0.2%
Synonym Absolute Ethanol
Solubility Information 1g/L in water
Molecular Weight (g/mol) 46.07
Percent Purity ≥99.5%
Grade BAKER ANALYZED™ HPLC
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol BAKER HPLC ANALYZED
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 99.5%, BAKER HPLC ANALYZED, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.