missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, for HPLC, Fisher Chemical™
C2H6O, CAS Number-64-17-5, alcohol, algrain, alcohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702
112.00 € - 264.00 €
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
MDL Number | 3568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10542382
|
Fisher Chemical
E/0665DF/15 |
1 L | Amber Glass Bottle |
112.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12438760
|
Fisher Chemical
E/0665DF/P17 |
2.5 L | Plastic Bottle |
264.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C2H6O, CAS Number-64-17-5, alcohol, algrain, alcohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702
Specifications
7 | |
64-17-5 | |
78°C | |
C2H6O | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 |
-114°C | |
Colorless | |
Liquid | |
3568 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol, absolute
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethanol Absolute, for HPLC, Fisher Chemical™