missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss. p.a., absolute, ≥99.8% (GC), for license holders only, Honeywell Riedel-de Haën™
99.99 € - 1394.02 €
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15643720
|
Honeywell-Riedel-de Haen
32221-2.5L-GL |
2.5 L | Glass Bottle |
153.36 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15633720
|
Honeywell-Riedel-de Haen
32221-2.5L |
2.5 L | Plastic Bottle |
141.36 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15653720
|
Honeywell-Riedel-de Haen
32221-2.5L-PC |
2.5 L | Poly-coated Bottle | N/A | |||||
15693720
|
Honeywell-Riedel-de Haen
32221-5L |
5 L | Plastic Bottle |
760.26 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15603730
|
Honeywell-Riedel-de Haen
32221-6X1L |
6 x 1 L | Plastic Bottle |
1068.35 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15673720
|
Honeywell-Riedel-de Haen
32221-4X2.5L |
4 x 2.5 L | Plastic Bottle |
1394.02 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15603720
|
Honeywell-Riedel-de Haen
32221-10L |
10 L | Stainless Steel Can | N/A | |||||
15663720
|
Honeywell-Riedel-de Haen
32221-30KG |
30 kg | Metal Drum |
408.43 €
30kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15613720
|
Honeywell-Riedel-de Haen
32221-160KG |
160 kg | Metal Drum | N/A | |||||
Description
Chemical Identifiers
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
64-17-5 | |
19% (60°F) | |
683°F | |
CH3CH2OH | |
44.6mmHg (20°C) | |
1718733 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
puriss. p.a. |
0.789g/mL at 25°C | |
14°C (57.2°F) | |
C2H6O | |
MFCD00003568 | |
1170 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
≥99.8% (GC) | |
Ethanol |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319
Danger
missing translation for 'einecsNumber' : 200-578-6
missing translation for 'rtecsNumber' : KQ6300000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.