missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-[3,5-di(trifluoromethyl)phenyl]-3-oxopropanoate, 90%, Thermo Scientific™
64.40 € - 470.00 €
Chemical Identifiers
CAS | 175278-02-1 |
---|---|
Molecular Formula | C13H10F6O3 |
Molecular Weight (g/mol) | 328.21 |
MDL Number | MFCD00052321 |
InChI Key | IBRYPSPFMRZTCX-UHFFFAOYSA-N |
Synonym | ethyl 3-3,5-bis trifluoromethyl phenyl-3-oxopropanoate, ethyl 3,5-bis trifluoromethyl benzoyl acetate, ethyl 3,5-bis trifluoromethyl benzoyl-acetate, ethyl 3-3,5-di trifluoromethyl phenyl-3-oxopropanoate, benzenepropanoic acid, b-oxo-3,5-bis trifluoromethyl-,ethyl ester |
PubChem CID | 2782049 |
IUPAC Name | ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate |
SMILES | CCOC(=O)CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10663423
|
Thermo Scientific Maybridge
TL00364DA |
1 g | Amber Glass Bottle |
64.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532473
|
Thermo Scientific Maybridge
TL00364EA |
10 g | Amber Glass Bottle |
470.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175278-02-1 | |
328.21 | |
IBRYPSPFMRZTCX-UHFFFAOYSA-N | |
2782049 | |
CCOC(=O)CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
C13H10F6O3 | |
MFCD00052321 | |
ethyl 3-3,5-bis trifluoromethyl phenyl-3-oxopropanoate, ethyl 3,5-bis trifluoromethyl benzoyl acetate, ethyl 3,5-bis trifluoromethyl benzoyl-acetate, ethyl 3-3,5-di trifluoromethyl phenyl-3-oxopropanoate, benzenepropanoic acid, b-oxo-3,5-bis trifluoromethyl-,ethyl ester | |
ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate |
Specifications
175278-02-1 | |
90% | |
(CF3)2C6H3C(O)CH2CO2CH2CH3 | |
ethyl 3-3,5-bis trifluoromethyl phenyl-3-oxopropanoate, ethyl 3,5-bis trifluoromethyl benzoyl acetate, ethyl 3,5-bis trifluoromethyl benzoyl-acetate, ethyl 3-3,5-di trifluoromethyl phenyl-3-oxopropanoate, benzenepropanoic acid, b-oxo-3,5-bis trifluoromethyl-,ethyl ester | |
CCOC(=O)CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F | |
328.21 | |
328.21 | |
Ethyl 3-[3, 5-di(trifluoromethyl)phenyl]-3-oxopropanoate |
90.0 | |
C13H10F6O3 | |
MFCD00052321 | |
IBRYPSPFMRZTCX-UHFFFAOYSA-N | |
ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate | |
2782049 | |
90% |