missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-aminobenzo[b]furan-2-carboxylate, 97%, Thermo Scientific Chemicals Product Code.: 11476220

Ethyl 3-aminobenzo[b]furan-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 11476220
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11476220

Brand: Thermo Scientific Alfa Aesar H27124.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39786-35-1
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00641330
InChI Key PWOARNMOPCOJEV-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzofuran-2-carboxylate, 3-aminocoumarilic acid ethyl ester, 2-benzofurancarboxylic acid, 3-amino-, ethyl ester, ethyl 3-aminobenzo b furan-2-carboxylate, ethyl 3-azanyl-1-benzofuran-2-carboxylate, 3-amino-2-benzofurancarboxylic acid ethyl ester, 3-aminobenzofuran-2-carboxylic acid ethyl ester, enamine_001390, ethyl 3-aminocoumarilate, ethyl3-aminobenzofuran-2-carboxylateShow MoreShow Less
PubChem CID 2063537
IUPAC Name ethyl 3-amino-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC=C2O1)N

Specifications

Melting Point 77°C to 82°C
Quantity 1 g
Formula Weight 205.22
Percent Purity 97%
Chemical Name or Material Ethyl 3-aminobenzo[b]furan-2-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10135422 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberH27124.06,H27124.03
Lot Number10135422 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27124.06,H27124.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-aminobenzo[b]furan-2-carboxylate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.