missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals

48.54 € - 387.00 €

Chemical Identifiers

CAS 539-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000251
InChI Key FQTIYMRSUOADDK-UHFFFAOYSA-N
Synonym ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
PubChem CID 68320
IUPAC Name ethyl 3-bromopropanoate
SMILES CCOC(=O)CCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10349053
View Documents
Thermo Scientific Acros
409810250
25 g
48.54 €
25g
Please to purchase this item. Need a web account? Register with us today!
10299550
View Documents
Thermo Scientific Acros
409811000
100 g
134.67 €
100g
Please to purchase this item. Need a web account? Register with us today!
10731434
View Documents
Thermo Scientific Acros
409815000
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

539-74-2
181.03
FQTIYMRSUOADDK-UHFFFAOYSA-N
68320
CCOC(=O)CCBr
C5H9BrO2
MFCD00000251
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
ethyl 3-bromopropanoate

Specifications

539-74-2
1.4190g/mL
70°C
98.5% min. (GC)
C5H9BrO2
1.4510 to 1.453
02, 256
1.419
FQTIYMRSUOADDK-UHFFFAOYSA-N
ethyl 3-bromopropanoate
68320
99%
Ethyl 3-bromopropionate, 99%
Colorless to Yellow
69°C to 71°C (10.0 mmHg)
Authentic
Glass Bottle
CH2BrCH2COOC2H5
MFCD00000251
15, 1444
ethyl 3-bromopropionate, propanoic acid, 3-bromo-, ethyl ester, 3-bromopropionic acid ethyl ester, ethyl beta-bromopropionate, unii-9b28g9s1jv, ccris 9051, propionic acid, 3-bromo-, ethyl ester, ethyl .beta.-bromopropionate, 3-bromo-propionic acid ethyl ester, ethyl-3-bromopropionate
CCOC(=O)CCBr
181.03
181.03
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-724-0

missing translation for 'tsca' : TSCA

Product Identifier
  • Ethyl 3-bromopropionate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.