missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4,4-difluorocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals
46.00 € - 500.00 €
Chemical Identifiers
CAS | 178312-47-5 |
---|---|
Molecular Formula | C9H14F2O2 |
Molecular Weight (g/mol) | 192.206 |
MDL Number | MFCD03788494 |
InChI Key | HZZDWLBBNSDYQM-UHFFFAOYSA-N |
PubChem CID | 2779138 |
IUPAC Name | ethyl 4,4-difluorocyclohexane-1-carboxylate |
SMILES | CCOC(=O)C1CCC(CC1)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369969
|
Thermo Scientific Alfa Aesar
H25754.MD |
250 mg | N/A | ||||||
11379969
|
Thermo Scientific Alfa Aesar
H25754.03 |
1 g |
148.74 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389969
|
Thermo Scientific Alfa Aesar
H25754.06 |
5 g |
500.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
178312-47-5 | |
192.206 | |
HZZDWLBBNSDYQM-UHFFFAOYSA-N | |
ethyl 4,4-difluorocyclohexane-1-carboxylate |
C9H14F2O2 | |
MFCD03788494 | |
2779138 | |
CCOC(=O)C1CCC(CC1)(F)F |
Specifications
178312-47-5 | |
C9H14F2O2 | |
ethyl 4,4-difluorocyclohexanecarboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester, ethyl4,4-difluorocyclohexanecarboxylate, 4,4-difluoro-1-ethoxycarbonyl cyclohexane, 4,4-difluoro-cyclohexanecarboxylic acid ethyl ester, ethyl 4,4-difluorocyclohexane carboxylate, ethyl-4,4-difluorocyclohexane carboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester 9ci, acmc-209xsu, 4-carbethoxy-1,1-difluorocyclohexane | |
CCOC(=O)C1CCC(CC1)(F)F | |
192.206 | |
192.21 | |
Ethyl 4,4-difluorocyclohexanecarboxylate |
84°C to 86°C (8 mmHg) | |
MFCD03788494 | |
HZZDWLBBNSDYQM-UHFFFAOYSA-N | |
ethyl 4,4-difluorocyclohexane-1-carboxylate | |
2779138 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title