missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals Product Code.: 11435543

Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals

Product Code. 11435543
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11435543

Brand: Thermo Scientific Alfa Aesar A13470.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

Solubility
Immiscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2969-81-5
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000259
InChI Key XBPOBCXHALHJFP-UHFFFAOYSA-N
Synonym ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl esterShow MoreShow Less
PubChem CID 76300
IUPAC Name ethyl 4-bromobutanoate
SMILES CCOC(=O)CCCBr

Specifications

Density 1.363
Boiling Point 80°C to 82°C (10 mm)
Flash Point 90°C (194°F)
Assay Percent Range 98%
Refractive Index 1.456
Quantity 50 g
Beilstein 1749700
Solubility Information Immiscible with water.
Formula Weight 195.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4-bromobutyrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10232205 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13470.30,A13470.0B,A13470.18
Lot Number10229495 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13470.30,A13470.0B,A13470.18
Lot NumberH7287A Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13470.30,A13470.0B,A13470.18
Lot Number10207536 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13470.30,A13470.0B,A13470.18
Lot Number10225381 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13470.30,A13470.0B,A13470.18
compliance-icons
Product Identifier
  • Ethyl 4-bromobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl 4-bromobutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.