Learn More
Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals
45.63 € - 463.31 €
Chemical Identifiers
CAS | 2969-81-5 |
---|---|
Molecular Formula | C6H11BrO2 |
Molecular Weight (g/mol) | 195.056 |
MDL Number | MFCD00000259 |
InChI Key | XBPOBCXHALHJFP-UHFFFAOYSA-N |
Synonym | ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester |
PubChem CID | 76300 |
IUPAC Name | ethyl 4-bromobutanoate |
SMILES | CCOC(=O)CCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435543
|
Thermo Scientific Alfa Aesar
A13470.18 |
50 g |
45.63 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445543
|
Thermo Scientific Alfa Aesar
A13470.30 |
250 g |
148.74 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425543
|
Thermo Scientific Alfa Aesar
A13470.0B |
1000 g |
463.31 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.
Solubility
Immiscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Chemical Identifiers
2969-81-5 | |
195.056 | |
XBPOBCXHALHJFP-UHFFFAOYSA-N | |
76300 | |
CCOC(=O)CCCBr |
C6H11BrO2 | |
MFCD00000259 | |
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester | |
ethyl 4-bromobutanoate |
Specifications
2969-81-5 | |
80°C to 82°C (10 mm) | |
98% | |
1.456 | |
1749700 | |
Immiscible with water. | |
CCOC(=O)CCCBr | |
195.056 | |
195.06 | |
Liquid |
1.363 | |
90°C (194°F) | |
C6H11BrO2 | |
MFCD00000259 | |
ethyl 4-bromobutyrate, 4-bromobutyric acid ethyl ester, ethyl-4-bromobutyrate, butanoic acid, 4-bromo-, ethyl ester, ethyl-4-bromotbutyrate, 4-bromo-n-butyric acid ethyl ester, brch2ch2ch2c o oc2h5, 4-bromo-butyric acid ethyl ester, 4-bromo-butanoic acid ethyl ester, gamma-bromobutyric acid ethyl ester | |
XBPOBCXHALHJFP-UHFFFAOYSA-N | |
ethyl 4-bromobutanoate | |
76300 | |
98% | |
Ethyl 4-bromobutyrate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 221-005-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Ethyl 4-bromobutyrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Ethyl 4-bromobutyrate
RUO – Research Use Only