missing translation for 'onlineSavingsMsg'
Learn More
Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals Product Code.: 11499503

Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals

Product Code. 11499503
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11499503

Brand: Thermo Scientific Alfa Aesar A12366.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-05-3
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00009136
InChI Key KTMGNAIGXYODKQ-SREVYHEPSA-N
Synonym ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoateShow MoreShow Less
PubChem CID 1715183
IUPAC Name ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
SMILES CCO\C=C(\C#N)C(=O)OCC

Specifications

Melting Point 47°C to 52°C
Boiling Point 190°C to 191°C (30 mmHg)
Flash Point 130°C (266°F)
Quantity 50 g
UN Number UN3439
Beilstein 878814
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10216265 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog NumberA12366.30,A12366,A12366.0B,A12366.18
Lot Number10155311 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12366.30,A12366.0B,A12366.18
Lot Number10104073 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12366.30,A12366.0B,A12366.18
Lot NumberGBFA025359 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12366.30,A12366.0B,A12366.18
Lot Number10211140 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12366.30,A12366.0B,A12366.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.