missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-(2,4-dinitrophenoxy)acetimidate, 98+%, Thermo Scientific™

59.70 € - 553.00 €

Chemical Identifiers

CAS 54322-32-6
Molecular Formula C10H11N3O6
Molecular Weight (g/mol) 269.213
MDL Number MFCD00010133
InChI Key UVBZLMGBNXCYNA-YRNVUSSQSA-N
Synonym ethyl n-2,4-dinitrophenoxy acetimidate, pubchem12897, jlyppahrtackn bt jw dtttrrvbapemaadmu@dmp, ethyl e-n-2,4-dinitrophenoxy acetimidate, 1-1e-2-ethoxy-1-azaprop-1-enyloxy-2,4-dinitrobenzene
PubChem CID 9566065
IUPAC Name ethyl (1E)-N-(2,4-dinitrophenoxy)ethanimidate
SMILES CCOC(=NOC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491230
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26341.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11401240
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26341.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11411240
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26341.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

54322-32-6
269.213
UVBZLMGBNXCYNA-YRNVUSSQSA-N
9566065
CCOC(=NOC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C
C10H11N3O6
MFCD00010133
ethyl n-2,4-dinitrophenoxy acetimidate, pubchem12897, jlyppahrtackn bt jw dtttrrvbapemaadmu@dmp, ethyl e-n-2,4-dinitrophenoxy acetimidate, 1-1e-2-ethoxy-1-azaprop-1-enyloxy-2,4-dinitrobenzene
ethyl (1E)-N-(2,4-dinitrophenoxy)ethanimidate

Specifications

54322-32-6
C10H11N3O6
Moisture sensitive
UVBZLMGBNXCYNA-YRNVUSSQSA-N
ethyl (1E)-N-(2,4-dinitrophenoxy)ethanimidate
9566065
≥98%
110°C to 113°C
MFCD00010133
ethyl n-2,4-dinitrophenoxy acetimidate, pubchem12897, jlyppahrtackn bt jw dtttrrvbapemaadmu@dmp, ethyl e-n-2,4-dinitrophenoxy acetimidate, 1-1e-2-ethoxy-1-azaprop-1-enyloxy-2,4-dinitrobenzene
CCOC(=NOC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])C
269.213
269.21
Ethyl N-(2,4-dinitrophenoxy)acetimidate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications