missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate, 97%, Thermo Scientific™
12.90 € - 208.00 €
Chemical Identifiers
CAS | 61713-23-3 |
---|---|
Molecular Formula | C9H16O4S2 |
Molecular Weight (g/mol) | 252.343 |
MDL Number | MFCD00026901 |
InChI Key | NBNGVIRHOBZBHM-UHFFFAOYSA-N |
Synonym | methylenebis ethyl thioglycolate, ethyl 2-2-ethoxy-2-oxoethyl thio methyl thio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, ethyl 2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, diethyl 2,2'-methylenedisulfanediyl diacetate, diethyl 2,2'-methylenebis sulfanediyl diacetate, ethyl 2-ethoxycarbonyl methylthio methylthio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanylmethylsulfanyl acetate, aceticacid, 2,2'-methylenebis thio bis-, diethyl ester 9ci |
PubChem CID | 521987 |
IUPAC Name | ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanylmethylsulfanyl]acetate |
SMILES | CCOC(=O)CSCSCC(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10223092
|
Thermo Scientific Maybridge
S12083DA |
1 g |
12.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10485712
|
Thermo Scientific Maybridge
S12083EA |
10 g |
69.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10190602
|
Thermo Scientific Maybridge
S12083EE |
50 g |
208.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
61713-23-3 | |
252.343 | |
NBNGVIRHOBZBHM-UHFFFAOYSA-N | |
521987 | |
CCOC(=O)CSCSCC(=O)OCC |
C9H16O4S2 | |
MFCD00026901 | |
methylenebis ethyl thioglycolate, ethyl 2-2-ethoxy-2-oxoethyl thio methyl thio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, ethyl 2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, diethyl 2,2'-methylenedisulfanediyl diacetate, diethyl 2,2'-methylenebis sulfanediyl diacetate, ethyl 2-ethoxycarbonyl methylthio methylthio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanylmethylsulfanyl acetate, aceticacid, 2,2'-methylenebis thio bis-, diethyl ester 9ci | |
ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanylmethylsulfanyl]acetate |
Specifications
61713-23-3 | |
97% | |
C9H16O4S2 | |
MFCD00026901 | |
NBNGVIRHOBZBHM-UHFFFAOYSA-N | |
ethyl 2-[(2-ethoxy-2-oxoethyl)sulfanylmethylsulfanyl]acetate | |
521987 | |
97% |
97.0 | |
Amber glass bottle | |
CH3CH2O2CCH2SCH2SCH2CO2CH2CH3 | |
methylenebis ethyl thioglycolate, ethyl 2-2-ethoxy-2-oxoethyl thio methyl thio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, ethyl 2-ethoxy-2-oxoethyl sulfanyl methyl sulfanyl acetate, diethyl 2,2'-methylenedisulfanediyl diacetate, diethyl 2,2'-methylenebis sulfanediyl diacetate, ethyl 2-ethoxycarbonyl methylthio methylthio acetate, ethyl 2-2-ethoxy-2-oxoethyl sulfanylmethylsulfanyl acetate, aceticacid, 2,2'-methylenebis thio bis-, diethyl ester 9ci | |
CCOC(=O)CSCSCC(=O)OCC | |
252.343 | |
252.35 | |
Ethyl 2-({[(2-ethoxy-2-oxoethyl)thio]methyl}thio)acetate |