Learn More
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
74.42 € - 823.10 €
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
MDL Number | MFCD00002885 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
10326740
|
Thermo Scientific Acros
146750010 |
1 L | Glass Bottle |
74.42 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10695572
|
Thermo Scientific Acros
146750025 |
2.5 L | Glass Bottle |
142.71 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10336740
|
Thermo Scientific Acros
146750250 |
25 L | Plastic Drum |
823.10 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
MFCD00002885 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
Specifica
-13°C | |
>500°C | |
196.0°C to 198.0°C | |
Authentic | |
1.4300 to 1.4330 (20°C, 589nm) | |
C2H6O2 | |
MFCD00002885 | |
01, 465 | |
01,375; 05,296; 09,217; 15,156 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
≥99.0% | |
Ethylene glycol, 99+% |
107-21-1 | |
1.1130g/mL | |
116°C (240.8°F) | |
Viscous Liquid | |
99+% | |
HOCH2CH2OH | |
Vapor Pressure: 0.12mmHg at 20°C | |
0.1% max. (Coulometric) | |
15, 3852 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.06 | |
Extra Pure |
Sicurezza e movimentazione
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 203-473-3
missing translation for 'rtecsNumber' : KW2975000
missing translation for 'tsca' : TSCA
- Ethylene glycol
Avvertenza
- Warning
Categoria di pericolo
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Indicazioni di pericolo
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
Consigli di prudenza
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.