missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

149.00 € - 495.00 €

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15598564
View Documents Promotion Details
Avantor J.T.Baker
9300-01
500 mL Narrow Mouth Amber Glass Bottle
149.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15508574
View Documents Promotion Details
Avantor J.T.Baker
9300-03
4 L Narrow Mouth Amber Glass Bottle
495.00 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Laboratory,Research,or Manufacturing Use
Specifications

Chemical Identifiers

107-21-1
62.068
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
C2H6O2
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol

Specifications

107-21-1
1L = 1.10kg
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
BAKER ANALYZED™ Reagent
111.00°C
C2H6O2
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07
Ethylene Glycol
Videos
SDS
Documents

Documents

Product Certifications