Learn More
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
67.23 € - 1487.40 €
Specifications
Chemical Name or Material | Ethylene glycol dimethyl ether |
---|---|
pH | 6 to 7.5 (1:9) |
Melting Point | -69.0°C |
Name Note | Stabilized with BHT |
CAS | 128-37-0 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10396550
|
Thermo Scientific
118455000 |
500 mL | Glass Bottle |
67.23 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696682
|
Thermo Scientific Acros
118450010 |
1 L | Glass Bottle |
113.57 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606692
|
Thermo Scientific Acros
118450025 |
2.5 L | Glass Bottle |
218.09 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247752
|
Thermo Scientific Acros
118450250 |
25 L | Metal Drum |
1487.40 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethylene glycol dimethyl ether | |
-69.0°C | |
128-37-0 | |
84.0°C to 86.0°C | |
Authentic | |
1.3785 to 1.3805 | |
0.867 | |
1.1 mPa.s (20°C) | |
0.1% max. (K.F.) | |
15, 3245 | |
90.12 | |
Extra Pure |
6 to 7.5 (1:9) | |
Stabilized with BHT | |
0.8670g/mL | |
−6°C | |
Liquid | |
99+% | |
CH3OCH2CH2OCH3 | |
01, 467 | |
01,267; 02,143; 16,229 | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
≥99% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May damage fertility. May damage the unborn child.
Highly flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Ethylene glycol dimethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only