Learn More
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 118450250
1480.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Ethylene glycol dimethyl ether | |
-69.0°C | |
128-37-0 | |
84.0°C to 86.0°C | |
−6°C | |
Metal Drum | |
1.3785 to 1.3805 | |
0.867 | |
1.1 mPa.s (20°C) | |
0.1% max. (K.F.) | |
15, 3245 | |
90.12 | |
Extra Pure |
6 to 7.5 (1:9) | |
Stabilized with BHT | |
0.8670g/mL | |
25 L | |
Authentic | |
Liquid | |
99+% | |
CH3OCH2CH2OCH3 | |
01, 467 | |
01,267; 02,143; 16,229 | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
≥99% |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Ethylene glycol dimethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0454286 | Certificate of Analysis | 31/01/2025 | 118450025,118450010,118450250,118455000 |
A0466238 | Certificate of Analysis | 24/09/2024 | 118450025,118450010,118450250,118455000 |
A0454286 | Certificate of Analysis | 25/08/2023 | 118450025,118450010,118450250,118455000 |
A0399540 | Certificate of Analysis | 15/12/2022 | 118450025,118450010,118450250,118455000 |
A0399541 | Certificate of Analysis | 15/12/2022 | 118450025,118450010,118450250,118455000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only