Learn More
Ethylenediamine, 99+%, Thermo Scientific Chemicals
34.07 € - 205.02 €
Chemical Identifiers
CAS | 107-15-3 |
---|---|
Molecular Formula | C2H8N2 |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00008204 |
InChI Key | PIICEJLVQHRZGT-UHFFFAOYSA-N |
Synonym | ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin |
PubChem CID | 3301 |
ChEBI | CHEBI:30347 |
IUPAC Name | ethane-1,2-diamine |
SMILES | NCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10593191
|
Thermo Scientific
220422500 |
250 mL | Glass bottle |
34.07 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328310
|
Thermo Scientific Acros
220420010 |
1 L | Glass bottle |
93.06 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10514474
|
Thermo Scientific Acros
220420025 |
2.5 L | Glass bottle |
205.02 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-15-3 | |
60.10 | |
PIICEJLVQHRZGT-UHFFFAOYSA-N | |
3301 | |
ethane-1,2-diamine |
C2H8N2 | |
MFCD00008204 | |
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
CHEBI:30347 | |
NCCN |
Specifications
107-15-3 | |
0.8980g/mL | |
38°C | |
99% min. (GC) | |
H2NCH2CH2NH2 | |
MFCD00008204 | |
01,372; 04,231; 13,157 | |
0.898 | |
Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol, slightly soluble in ether, soluble in benzene | |
NCCN | |
60.10 | |
CHEBI:30347 | |
60.1 | |
Extra Pure | |
Ethylenediamine |
11.0°C | |
117.0°C to 118.0°C | |
Authentic | |
C2H8N2 | |
1.4555 to 1.4575 | |
04, 230 | |
15, 3849 | |
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin | |
PIICEJLVQHRZGT-UHFFFAOYSA-N | |
ethane-1,2-diamine | |
3301 | |
1.6 mPa.s (20°C) | |
99+% | |
Liquid, Fuming In Air |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-468-6
- Ethylenediamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.