missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
25.25 € - 416.00 €
Chemical Identifiers
CAS | 60-00-4, 7732-18-5 |
---|---|
Molecular Formula | C10H16N2O8 |
Molecular Weight (g/mol) | 292.24 |
MDL Number | MFCD00003541 |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
Synonym | edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10306983
|
Fisher BioReagents
BP2482-100 |
100 mL | Poly Bottle |
25.25 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10182903
|
Fisher BioReagents
BP2482-500 |
500 mL | Poly Bottle |
37.90 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10628203
|
Fisher BioReagents
BP2482-1 |
1 L | Nalgene Poly Bottle |
61.05 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073074
|
Fisher BioReagents
BP2482-20 |
20 L | Poly CUBE |
416.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.Chemical Identifiers
60-00-4, 7732-18-5 | |
292.24 | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
6049 | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
C10H16N2O8 | |
MFCD00003541 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
CHEBI:42191 | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
Colorless | |
C10H16N2O8 | |
MFCD00003541 | |
15, 3565 | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid | |
6049 | |
292.25 | |
8.0 | |
Ethylenediamine Tetraacetic Acid, 0.5 M Solution, pH 8.0 (for DNA Work) |
60-00-4, 7732-18-5 | |
(HOOCCH2)2NCH2CH2N(CH2COOH)2 | |
0.475 to 0.525M | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
292.24 | |
CHEBI:42191 | |
Pass Test | |
Liquid |
Safety and Handling
Recommended Storage : RT
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.