missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Biological buffer, Chelation/Complexation reagent
33.00 € - 604.00 €
Chemical Identifiers
CAS | 6381-92-6 |
---|---|
Molecular Formula | C10H18N2Na2O10 |
Molecular Weight (g/mol) | 372.24 |
MDL Number | MFCD00150037,MFCD00003541 |
InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
PubChem CID | 44120005 |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10031660
|
Thermo Scientific Chemicals
147851000 |
100 g | Plastic bottle |
33.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10416790
|
Thermo Scientific Chemicals
147855000 |
500 g | Plastic bottle |
98.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10706351
|
Thermo Scientific Chemicals
147850010 |
1 kg | Plastic bottle |
167.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10524634
|
Thermo Scientific Chemicals
147850050 |
5 kg | Plastic drum |
604.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Chelation/Complexation reagentChemical Identifiers
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Specifications
6381-92-6 | |
White | |
11.4% max. (200°C, 3 hrs) | |
99.0 to 101% (On dry substance) | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate | |
44120005 | |
99+% | |
Fine Crystalline Powder |
252.0°C | |
4.0 to 6.0 (1% soln. at 20°C) | |
Authentic | |
C10H18N2Na2O10 | |
15, 3565 | |
Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
372.23 | |
Analysis | |
Ethylenediaminetetraacetic acid, disodium salt dihydrate |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethylenediaminetetraacetic acid, disodium salt dihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only