Learn More
Ethynyl Estradiol, MedChemExpress
MedChemExpress Ethynyl Estradiol (17α-Ethynylestradiol;Ethynylestradiol) is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills.
Brand:  Medchem Express HY-B0216
Encompass_Preferred
 View more versions of this product
Product Code. 16432066
  • 41.85 € / 500mg
Explore more special offers
Description
Description
Ethynyl Estradiol (17α-Ethynylestradiol
Specifications
Specifications
Ethynyl Estradiol
White
C20H24O2
17α-Ethynylestradiol Ethynylestradiol
OC1=CC=C2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(C#C)O)=C1
296.4
Research
Cancer-programmed cell death
57-63-6
Research
500 mg
DMSO : 250 mg/mL (843.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
296.4
99.28%
Solid