missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl Estradiol, MedChemExpress

MedChemExpress Ethynyl Estradiol (17α-Ethynylestradiol;Ethynylestradiol) is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills.

52.65 € - 86.40 €

Specifications

Chemical Name or Material Ethynyl Estradiol
CAS 57-63-6
Color White
Purity Grade Notes Research
Molecular Formula C20H24O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16458176
Encompass_Preferred
View Documents
Medchem Express
HY-B0216
100 mg
52.65 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16432066
Encompass_Preferred
View Documents
Medchem Express
HY-B0216
500 mg
86.40 €
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethynyl Estradiol (17α-Ethynylestradiol
Specifications

Specifications

Ethynyl Estradiol
White
C20H24O2
DMSO : 250 mg/mL (843.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
296.4
99.28%
Solid
57-63-6
Research
17α-Ethynylestradiol Ethynylestradiol
OC1=CC=C2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(C#C)O)=C1
296.4
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

H302∣H350

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only