missing translation for 'onlineSavingsMsg'
Learn More
Etoposide, Thermo Scientific™ Product Code.: 15405829

Etoposide, Thermo Scientific™

Product Code. 15405829
100mg
Click to view available options
Quantity:
25mg
100mg
This item is not returnable. View return policy

Product Code. 15405829

Brand: Thermo Scientific Alfa Aesar J63651.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Topoisomerase inhibitor and inducer of apoptosis

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Etoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.

Solubility
Insoluble in water.

Notes
Store solutions in DMSO at 4°C. For long term storage prepare aliquots and store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33419-42-0
Molecular Formula C29H32O13
Molecular Weight (g/mol) 588.56
MDL Number MFCD00869325,MFCD00869325
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
Synonym vjjpusntgommgy-nzlmilqcsa
PubChem CID 50936917
IUPAC Name (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-oneShow MoreShow Less
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12Show MoreShow Less

Specifications

Quantity 100mg
Merck Index 14,3886
Solubility Information Insoluble in water.
Optical Rotation −107° (c=1 in Methanol/Dichloromethane 1:9)
Formula Weight 588.56
Chemical Name or Material Etoposide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ02L021 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberJ63651.MB,J63651.MC
Lot NumberQ02L020 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberJ63651.MB,J63651.MC
Lot NumberN11L021 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberJ63651.MB,J63651.MC
Lot NumberM31L027 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog NumberJ63651.MB,J63651.MC
Lot NumberY13K046 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ63651.MB,J63651.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Etoposide, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.