missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Podophyllotoxin, 95%
Click to view available options
Quantity:
100 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 178.0°C to 184.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 100 mg |
Merck Index | 15, 7663 |
Specific Rotation | − 110.70 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone and acetic acid |
Formula Weight | 414.4 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462469 | Certificate of Analysis | 20/05/2024 | 228410010,228411000 |
A0426672 | Certificate of Analysis | 25/08/2023 | 228410010,228411000 |
A0374590 | Certificate of Analysis | 13/12/2022 | 228410010,228411000 |
A0406841 | Certificate of Analysis | 28/09/2021 | 228410010,228411000 |
Safety and Handling

Product Identifier
- Podophyllotoxin
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Podophyllotoxin, 95% >
Spot an opportunity for improvement?Share a Content Correction