missing translation for 'onlineSavingsMsg'
Learn More
Europium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11321039

Europium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11321039
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11321039

Brand: Thermo Scientific Alfa Aesar 011298.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuCl3·xH2O
MDL Number MFCD00010994

Specifications

Color Yellow
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 258.32 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) chloride hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251561 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011298.06,011298.14
Lot NumberI03U022 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011298.06,011298.14
Lot NumberK15Z042 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011298.06,011298.14
Lot NumberF12R047A Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011298.06,011298.14
Lot NumberU14F005 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011298.06,011298.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Europium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.