Learn More
Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
129.00 € - 1159.00 €
Chemical Identifiers
CAS | 10031-53-5 |
---|---|
Molecular Formula | EuH12N3O15 |
Molecular Weight (g/mol) | 446.066 |
MDL Number | MFCD00149700 |
InChI Key | JVYYYCWKSSSCEI-UHFFFAOYSA-N |
Synonym | europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g |
PubChem CID | 202256 |
IUPAC Name | europium(3+);trinitrate;hexahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387108
|
Thermo Scientific Alfa Aesar
015290.04 |
2 g |
129.00 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397108
|
Thermo Scientific Alfa Aesar
015290.09 |
10 g |
312.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480480
|
Thermo Scientific Alfa Aesar
015290.18 |
50 g |
1159.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Europium(III) nitrate hexahydrate is used in the preparation of pharmaceuticals, agrochemicals and other chemical products. It is used as a catalyst and a component of specialized steels. It is also used in the synthesis of light emitting nanomaterials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10031-53-5 | |
446.066 | |
JVYYYCWKSSSCEI-UHFFFAOYSA-N | |
202256 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3] |
EuH12N3O15 | |
MFCD00149700 | |
europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g | |
europium(3+);trinitrate;hexahydrate |
Specifications
85°C | |
Colorless | |
99.9% (REO) | |
MFCD00149700 | |
Soluble in water. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3] | |
446.066 | |
446.06 (337.97 Anhydrous) | |
Colorless crystalline |
10031-53-5 | |
Crystalline or Powder | |
EuH12N3O15 | |
europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g | |
JVYYYCWKSSSCEI-UHFFFAOYSA-N | |
europium(3+);trinitrate;hexahydrate | |
202256 | |
Odorless | |
Europium(III) nitrate hexahydrate |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c
H272-H315-H319-X
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 233-380-3
missing translation for 'rtecsNumber' : LE8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only