Learn More
Thermo Scientific™ Eutech™ pH 6+ pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
483.00 € - 536.67 €
Specifications
Accuracy (mV) | ±1 |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.05°C |
Range (pH) | 0 to 14 |
Range (mV) | -1000 to 1000 |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15716275
|
Thermo Scientific™
01X245026 |
Eutech pH 6+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each), Rubber Armour/Stand | N/A | N/A | N/A | |||||
10548135
|
Thermo Scientific™
01X245026 |
Eutech pH 6+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each), Rubber Armour/Stand |
507.53 €
Each |
In Stock
Log in to see stock. |
||||||
11559164
|
Thermo Scientific™
01X245027 |
Eutech pH 6+ pH Meter with 3-in-1 pH/Temperature Combi Electrode |
536.67 €
Each |
In Stock
Log in to see stock. |
||||||
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 6+ pH Meter ECPH602PLUSK with single Junction pH electrode and ECPH603PLUSK with 3-in-1 pH/Temperature Combi Electrode are ideal choices for routine applications in laboratories, productions plants and schools.
Features:
- multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Hold Functionmomentarily freezes reading for easy viewing and recording
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic withmessage codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shieldsmeter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-sitemeter calibration andmeasurement
- pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST),4.10, 6.97 (Pb)
Specifications
±1 | |
±0.05°C | |
-1000 to 1000 | |
Up to 5 | |
0.1°C | |
Automatic/Manual (0°C to 100°C) |
±0.01 | |
0 to 14 | |
1 mV | |
0°C to 100°C | |
LCD | |
4 AAA x 1.4V batteries |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.