Learn More
WTW™ Exchange Membrane Heads for Dissolved Oxygen Sensors
For use with DO sensors, these replacement heads for membrane-covered galvanic dissolved oxygen sensors, are ideal for measuring in water and waterous solutions.
Brand: WTW™ 202725
181.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Replacement head for membrane-covered galvanic dissolved oxygen sensors to measure in water and waterous solutions
- 3 Replacement membrane heads for all dissolved oxygen sensors, except StirrOx™G, DurOx™325
- For use with DO sensor
- Exchange membrane head type
Specifications
Dissolved Oxygen Sensor | |
Exchange Membrane Head |