Learn More
Novus Biologicals™ Exosome Standards (B16F10 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP2-49866-200ug
 View more versions of this product
Product Code. 18682591
  • 529.00 € / Pack of 2
Explore more special offers
Specifications
Store at 4°C. After reconstitution store at -70°C.
LYOPH
2 x 100 μg Vials