missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Exosome Standards (B16F10 cell line)

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

417.00 € - 1345.00 €

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18684351
View Documents
Novus Biologicals™
NBP2-49866-60ug
2 x 30 μg Vials
417.00 €
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
18650621
View Documents
Novus Biologicals™
NBP2-49866-180ug
6 x 30 μg Vials
1080.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
18682591
View Documents
Novus Biologicals™
NBP2-49866-200ug
2 x 100 μg Vials
529.00 €
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
18692650
View Documents
Novus Biologicals™
NBP2-49866-600ug
6 x 100 μg Vials
1345.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Store at 4°C. After reconstitution store at -70°C.
LYOPH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.