missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome Standards (COLO1 cell line)
Assay Kit (Colorimetric)
417.00 € - 1345.00 €
Specifications
Content And Storage | Store at 4°C. After reconstitution store at -70°C. |
---|---|
Concentration | LYOPH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18655380
|
Novus Biologicals™
NBP2-49845-180ug |
6 x 30 μg Vials |
1080.00 €
Pack of 6 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
18644351
|
Novus Biologicals™
NBP2-49845-200ug |
2 x 100 μg Vials |
543.00 €
Pack of 2 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
18652591
|
Novus Biologicals™
NBP2-49845-600ug |
6 x 100 μg Vials |
1345.00 €
Pack of 6 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
18694040
|
Novus Biologicals™
NBP2-49845-60ug |
2 x 30 μg Vials |
417.00 €
Pack of 2 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
Descripción
Especificaciones
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto