missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Expi293F™ Cells
Expi293F™ human cells are derived from the 293 cell line, and are a core component of the Expi293™ Expression System
Brand: Gibco™ A14528
This item is not returnable.
View return policy
Description
- Expi293F™ human cells are derived from the 293 cell line, and are a core component of the Expi293™ Expression System
- They are maintained in suspension culture and will grow to high density in Expi293™ Expression Medium
- Expi293F™ cells are highly transfectable and generate superior protein yields compared to standard 293 cell lines in transient protein expression
- Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293™ Expression Medium and 10% DMSO
- The cells should be thawed directly into Expi293™ Expression Medium
- The six-vial multipack eliminates the need to expand and rebank the cells in the lab
- Cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator
- However, cultures are scalable from volumes of 10 L in bioreactors
- Protein harvest can be performed at 2 to 7 days post-transfection, depending on the protein expressed
- For expression of recombinant antibodies, harvest is typically at 5 to 7 days post-transfection
- There is no need for culture medium supplementation or batch feeding during expression
Specifications
1 x 107 cells/mL, store in liquid nitrogen | |
Kidney (Embryonic) | |
Cryopreserved | |
Expi293™ Expression Medium | |
Expi293™ | |
6 x 1 mL |
Expi293F | |
Suspension Cell Culture | |
Human | |
1 x 107 | |
293-F Cells |
For Research Use Only. Not for use in diagnostic procedures.