Learn More
Gibco™ ExpiCHO™ Expression System Kit
The ExpiCHO Expression System Kit combines the power of rapid, ultra-high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells.
Brand:  Gibco™ A29133
3367.75 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
 View more versions of this product
Product Code. 15441923
  • 3725.00 € / 1 set
Explore more special offers
Includes: 2 vials frozen ExpiCHO-S™ cells, 1 bottle 1000mL ExpiCHO™ Expression Medium, 1 ExpiFectamine™ CHO transfection kit for 1 L of culture, 1 vial Antibody Expressing Positive Control Vector, 1 bottle 100mL OptiPRO™ SFM; store all components at +2° - +8° C, except frozen cells stored in liquid nitrogen
Description
Description
The ExpiCHO Expression System Kit combines the power of rapid, ultra–high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells. It is based on high-density culture of ExpiCHO-S cells in ExpiCHO Expression Medium. Transient expression is powered by ExpiFectamine CHO transfection reagent, designed specifically for high-density CHO suspension culture, in combination with a specialized transfection enhancer and culture feed.
All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.
The ExpiCHO Expression System enables:
  • Milligram to multigram yields of recombinant protein per liter of culture in 5–14 days post-transfection
  • Robust and reproducible transfection results, for greater confidence in results and ability to use CHO cells from research to discovery
  • Transfection efficiency of > 90% for high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Use of one-third to one-half less plasmid DNA than other transfection reagents 
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows
  • Scalable expression for culture formats from  <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks
The components included in the ExpiCHO Expression System Kit are: 2 vials of frozen ExpiCHO-S cells, 1 L of ExpiCHO Expression Medium, 1 ExpiFectamine CHO Transfection Kit sufficient to transfect 1 L of culture, OptiPRO SFM, and an antibody-expressing positive control vector.
ExpiCHO-S cells, ExpiCHO Expression Medium, ExpiFectamine CHO Transfection Kit, and OptiPRO SFM may also be purchased separately.
All components of the system are animal origin–free.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Expression System
Transfection
• 2 vials frozen ExpiCHO-S cells
• 1000 mL ExpiCHO Expression Medium
• 1 ExpiFectamine CHO Transfection Kit for 1 L of culture
• 1 vial Antibody Expressing Positive Control Vector
• 100 mL OptiPRO SFM
Store all components at 2-8°C, except frozen cells which should be stored in liquid nitrogen.
CHO-S, SFM Adapted
ExpiCHO™
CMV
Untagged
1 kit
Approved for shipment at Room Temperature or on Wet or Dry Ice
Mammalian
Protein Production, Constitutive Expression
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ ExpiCHO™ Expression System Kit > Cell Line: CHO-S, SFM Adapted