missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ GreenER™ EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
666.00 € - 2670.00 €
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Fast or Standard |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10022303
|
SYBR™ GreenER™
11794200 |
200 Reactions |
666.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10164233
|
SYBR™ GreenER™
1179401K |
1000 Reactions |
2670.00 €
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This EXPRESS SYBR GreenER qPCR SuperMix comes with premixed ROX reference dye.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
SYBR | |
qPCR | |
Fast or Standard | |
SYBR™ GreenER™ | |
Gene Expression |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
Real Time PCR SYBR Master Mix | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.