missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar
The durable ribbed high-strength aluminium body, and superior vacuum performance and insulation provide versatile extended-time storage.
Brand: Taylor Wharton™ 10XTB-11M
This item is not returnable.
View return policy
Description
- Provides extended-time storage
- High-strength aluminium body
- Provides maximum holding times with superior vacuum performnce and insulation
Specifications
Long-term Cyrogenice Storage Dewar | |
10 L | |
Cryogenic storage |
10XTB-11M | |
Aluminum | |
Six 11 in. canisters |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar > Capacity: 10 L
Spot an opportunity for improvement?Share a Content Correction