Learn More
Ismatec™ Extension Tubing, Tygon ST
For use with 2- and 3-stop color-coded tubing.
Brand:  Ismatec™ SC0042T
 View more versions of this product
Product Code. 15773319
  • 153.00 € / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inexpensive, all-around tubing for general laboratory applications.
  • Transparent
  • Resistant to almost all inorganic chemicals
  • Tasteless
  • Smooth, polished inner wall
  • Low gas permeability
  • Non-aging and non-oxidizing
  • High dialectric constant
Specifications
Specifications
FDA 21 CFR175.300
1.65 mm
10 m
−50°C
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky.Gas sterilization with Ethylene oxide.Not recommended for sterilization with radiation.
74°C
Autoclavable
Tygon ST R-3603, R-3607
General laboratory applications, 2- and 3-stop color-coded tubing.
1 Reel
I.D. 1.65 mm
Transparent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ismatec™ Extension Tubing, Tygon ST > I.D. 1.65 mm