missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Extractor acc. to Soxhlet

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 05367073

Product Code. 10634202

  • 1070.00 € / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Extractor head with flange LF 100
  • Made of DURAN™ tubing
  • Flat flange lid LF100 center neck NS 45/40
  • Clamp for LF100
  • Dimroth condenser with cone
  • With thread connectors GL 14
  • Round bottom flask capacity: 2,000 mL NS 29/32
Specifications

Specifications

2 L
Flange
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente Extractor acc. to Soxhlet > 2000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.