Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05400158
This item is not returnable.
View return policy
Description
- Extractor heads acc. to DIN 12602
- With cone NS 29/32
- Available with/without NS stopcock
- Made of DURAN™ tubing
Specifications
500 mL | |
Borosilicate Glass 3.3 | |
NS 60/46 |
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet > 500 mL
Spot an opportunity for improvement?Share a Content Correction