Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
88.00 € - 274.00 €
Specifications
Material Borosilicate Glass 3.3
View More Specs
Products 7
Product Code Brand Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
10047910
View Documents
Lenz Laborglasintrumente
05395146
200 mL NS 45/40
88.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10681702
View Documents
Lenz Laborglasintrumente
05400131
70 mL NS 34/35
164.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10151521
View Documents
Lenz Laborglasintrumente
05400137
100 mL NS 45/40
178.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10558421
View Documents
Lenz Laborglasintrumente
05400149
250 mL NS 45/40
197.00 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
10663511
View Documents
Lenz Laborglasintrumente
05395152
300 mL NS 60/46
194.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10444451
View Documents
Lenz Laborglasintrumente
05395170
1 L NS 70/50
251.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
10121571
View Documents
Lenz Laborglasintrumente
05400158
500 mL NS 60/46
274.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
Description
Description
  • Extractor heads acc. to DIN 12602
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
Borosilicate Glass 3.3
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet