Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05360025
 View more versions of this product
Product Code. 10382761
  • 287.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The method allows an effective separation with relatively small amounts of extracting solvents.
  • Extractor in compact and robust design, with flask connection NS 29/32
  • Made of DURAN™ tubing
  • Dimroth condenser with thread GL 14
  • Round bottom flask
Specifications
Specifications
100 mL
Duran Glass
NS 29/32