Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
287.00 € - 498.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10382761
|
Lenz Laborglasintrumente
05360025 |
100 mL |
287.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
10047850
|
Lenz Laborglasintrumente
05360043 |
150 mL |
344.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
10352811
|
Lenz Laborglasintrumente
05360058 |
500 mL |
498.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
10251511
|
Lenz Laborglasintrumente
05360031 |
70 mL |
300.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
10738841
|
Lenz Laborglasintrumente
05360052 |
300 mL |
490.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- The method allows an effective separation with relatively small amounts of extracting solvents.
- Extractor in compact and robust design, with flask connection NS 29/32
- Made of DURAN™ tubing
- Dimroth condenser with thread GL 14
- Round bottom flask
Specifications
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser