missing translation for 'onlineSavingsMsg'
Learn More

Eye Wash Station

Keep the Eye Wash Station Refill on hand per the 1997 Approved Code of PracticeP L74 regulation for contingency supplies in case of emergency.

Brand:  Safety First E481/

Product Code. 12980475

  • 63.86 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

The 1997 Approved Code of Practice states that Where tap water is not readily available for eye irrigation, at least 1L of sterile water or sterile normal saline 0.9% in sealed disposable containers should be provided.


Description

Description

  • Wall mountable
  • Integrated mirror
  • Two 500mL bottles
Specifications

Specifications

Two 500 mL Bottles and Mirror
Eyewash Station
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eye Wash Station >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.