Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
GREENER_CHOICE
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Brand:  Thermo Scientific™ 4641310N
315.99 EUR valid until 2025-12-31
Use promo code "24314" to get your promotional price.
 View more versions of this product
Product Code. 15976132
  • 350.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research.
Learn more >
Ergonomically In-touch 
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort 
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications. 
Wide Selection of ClipTip Pipette Tips 
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time Warranty
With convenient online registration, and may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ClipTip 12.5 Ext
Pink
F1-ClipTip™
0.1 to 2 μL
Pink
F1-ClipTip
1
Variable Volume Pipette
0.002μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products