Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
GREENER_CHOICE
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
289.44 € - 376.00 €
Specifications
Number of Channels 1
Product Line F1-ClipTip™
Type Variable Volume Pipette
View More Specs
Products 9
Product Code Brand Description Volume (Metric) Compatible Tips Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Compatible Tips Price Quantity & Availability  
15571895
View Documents
Thermo Scientific™
4641170N
F1-ClipTip 1 to 10 μL ClipTip 20 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15581895
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4641180N
F1-ClipTip 2 to 20 μL ClipTip 20
352.00 €
Each
In Stock
to see stock.
Add to basket
15591895
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4641190N
F1-ClipTip 5 to 50 μL ClipTip 50
352.00 €
Each
In Stock
to see stock.
Add to basket
15501905
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641200N
F1-ClipTip 10 to 100 μL ClipTip 200
352.00 €
Each
In Stock
to see stock.
Add to basket
15511905
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641210N
F1-ClipTip 20 to 200 μL ClipTip 200
376.00 €
Each
In Stock
to see stock.
Edge
Add to basket
15521905
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641220N
F1-ClipTip 30 to 300 μL ClipTip 300
352.00 €
Each
In Stock
to see stock.
Add to basket
15531905
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641230N
F1-ClipTip 100 to 1000 μL ClipTip 1000
356.00 €
Each
In Stock
to see stock.
Edge
Add to basket
15976132
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641310N
F1-ClipTip 0.1 to 2 μL ClipTip 12.5 Ext
350.00 €
Each
In Stock
to see stock.
Add to basket
15996132
GREENER_CHOICE
View accessory products missing translation for 'viewDocuments'
Thermo Scientific™
4641320N
F1-ClipTip 1 to 10 μL ClipTip 12.5 Ext
350.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research.
Learn more >
Ergonomically in-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure volume lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable comfort 
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications. 
Wide selection of ClipTip Pipette Tips 
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Lifetime warranty
With convenient online registration, and may vary by country.
Specifications
Specifications
1
F1-ClipTip™
Variable Volume Pipette