Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
Streamline daily pipetting with innovative interlocking technology, which ensures a complete seal with minimal tip attachment.
274.32 € - 275.40 €
Specifications
Certifications/Compliance The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
Model F1-ClipTip™
Number of Channels 1
Product Line F1-ClipTip™
Type Variable Volume Pipette
View More Specs
Products 7
Product Code Brand Volume (Metric) Compatible Tips Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Compatible Tips Color Price Quantity & Availability  
13246279
View Documents Promotion Details
Thermo Scientific™
4641170
1 to 10 μL F1-ClipTip 20 Pink This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
13256279
View Documents Promotion Details
Thermo Scientific™
4641180
2 to 20 μL F1-ClipTip 20 Pink This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
13266279
View Documents Promotion Details
Thermo Scientific™
4641190
5 to 50 μL F1-ClipTip 50 Violet This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
13276279
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
4641200
10 to 100 μL F1-ClipTip 200 Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
13286279
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
4641210
20 to 200 μL F1-ClipTip 200 Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
13206289
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
4641220
30 to 300 μL F1-ClipTip 300 Orange This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
13296279
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
4641230
100 to 1000 μL F1-ClipTip 1000 Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description
Description
Ensure a complete seal on every channel with minimal tip attachment and ejection force using Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes, which feature innovative ClipTip interlocking technology. Once attached, tips are locked firmly in place and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip pipette until it is released.
Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette
Ergonomic design
  • Offering low attachment, ejection and plunger forces for safe and comfortable use
Remarkably lightweight
  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
Adjustable comfort
  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
Super blow-out
  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
Soft-touch tip ejection
  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle push
Volume Lock Adjustment
  • The secure volume setting prevents undesirable volume drift during operation
Industry-leading lifetime warranty
  • With convenient online registration
Ordering Information:
† Available in 50-μL models and below
Compatible with:
Thermo Scientific ClipTip pipette tips.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
1
Variable Volume Pipette
F1-ClipTip™
F1-ClipTip™