missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bulk Beads
GREENER_CHOICE

For use with Bead Mill 4 and 24 Homogenizers

242.00 € - 322.00 €

Specifications

For Use With (Equipment) Bead Mill 4 and 24 Homogenizers
View More Specs

Products 3
Product Code Brand Particle Size Price Quantity & Availability  
Product Code Brand Particle Size Price Quantity & Availability  
15515809
GREENER_CHOICE
View Documents
Fisherbrand™
15515809
1.4 mm
322.00 €
1 pack
In Stock
to see stock.
Edge
Add to basket
 
15505809
View Documents
Fisherbrand™
15505809
2.38 mm
242.00 €
1 pack
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
15525809
GREENER_CHOICE
View Documents
Fisherbrand™
15525809
2.8 mm
251.00 €
1 pack
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Fill up to 500 x 2mL sample tubes
  • Sample tubes not included
Specifications

Specifications

Bead Mill 4 and 24 Homogenizers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Bulk Beads

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.