missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
12510007
|
Epredia™
9802 |
Xylene-free Clearant | N/A | N/A | N/A | |||||
Description
- Reduces Tissue processing time by 80%
- Decreases the amount of reagents used up to 75%
- Simplifies reagent management by eliminating the need to rotate reagents
- Compatible with H&E, IHC, Special Stains and In situ
Includes:
One 5L bottle each: Fixative (Solution 1), Dehydrant (Solution 2) and Clearing Reagent (Solution 3).
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Fast-Flex™ Processing Reagents